3,6-Dimethoxy-4-pyridazinecarboxylic acid ethyl ester ≥95%

Supplier: AOBChem USA

Warning

35596-500MG 35596-1G 35596-5G
77339-884EA 671.56 USD
77339-884 77349-554 77373-790
3,6-Dimethoxy-4-pyridazinecarboxylic acid ethyl ester ≥95%
3,6-Dimethoxy-4-pyridazinecarboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₄
MW: 212.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29072142
CAS Number: 262353-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR