3,6-Dimethoxy-4-pyridazinecarboxylic acid methyl ester ≥95%

Supplier: AOBChem USA

Warning

35595-500MG 35595-1G 35595-5G
77325-858EA 346.08 USD
77325-858 77349-552 77373-788
3,6-Dimethoxy-4-pyridazinecarboxylic acid methyl ester ≥95%
3,6-Dimethoxy-4-pyridazinecarboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₄
MW: 198.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11615773
CAS Number: 1097778-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR