3-Bromo-5-fluoro-4-hydroxyanisole ≥97%

Supplier: AOBChem USA

Warning

35587-500MG 35587-1G 35587-5G 35587-25G
77325-856EA 243.08 USD
77325-856 77349-548 77362-196 77385-188
3-Bromo-5-fluoro-4-hydroxyanisole ≥97%
3-Bromo-5-fluoro-4-hydroxyanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD28738845
CAS Number: 1782834-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR