2,3-Difluoro-4-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

35564-500MG 35564-1G 35564-5G
77336-586EA 389.34 USD
77336-586 77349-544 77373-780
2,3-Difluoro-4-(methylthio)benzoic acid ≥97%
2,3-Difluoro-4-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD32215269
CAS Number: 261915-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR