6-Bromo-2-chloro-3-hydroxy-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

35556-500MG 35556-1G 35556-5G
77337-554EA 421.27 USD
77337-554 77349-540 77373-776
6-Bromo-2-chloro-3-hydroxy-4-methylpyridine ≥97%
6-Bromo-2-chloro-3-hydroxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.467 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28099842
CAS Number: 1823888-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR