2-Bromo-6-fluoro-3-propoxyphenol ≥97%

Supplier: AOBChem USA

Warning

35548-500MG 35548-1G 35548-5G 35548-25G
77325-854EA 182.31 USD
77325-854 77346-928 77362-194 77385-186
2-Bromo-6-fluoro-3-propoxyphenol ≥97%
2-Bromo-6-fluoro-3-propoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD32631811
CAS Number: 2364585-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR