(2-Bromo-4,6-dichlorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

35544-500MG 35544-1G 35544-5G
77338-556EA 498.52 USD
77338-556 77349-538 77373-772
(2-Bromo-4,6-dichlorophenyl)methanol ≥97%
(2-Bromo-4,6-dichlorophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD27941813
CAS Number: 115615-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR