1-Bromo-3-(1-methylethyl)-2-(phenylmethoxy)-benzene ≥95%

Supplier: AOBChem USA

Warning

35526-500MG 35526-1G 35526-5G
77336-668EA 391.4 USD
77336-668 77349-536 77373-770
1-Bromo-3-(1-methylethyl)-2-(phenylmethoxy)-benzene ≥95%
1-Bromo-3-(1-methylethyl)-2-(phenylmethoxy)-benzene

Precaución: for Research Use Only

Formula: C₁₆H₁₇BrO
MW: 305.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32183356
CAS Number: 1637439-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR