4'-Methoxy-2-(trifluoromethyl)biphenyl-4-amine ≥97%

Supplier: AOBChem USA

Warning

35509-500MG 35509-1G 35509-5G 35509-25G
77325-850EA 184.37 USD
77325-850 77347-192 77362-190 77385-184
4'-Methoxy-2-(trifluoromethyl)biphenyl-4-amine ≥97%
4'-Methoxy-2-(trifluoromethyl)biphenyl-4-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₃NO
MW: 267.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13152300
CAS Number: 1044209-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR