5-Fluoro-2-propoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

35493-500MG 35493-1G 35493-5G 35493-25G
77325-844EA 147.29 USD
77325-844 77345-256 77362-184 77385-178
5-Fluoro-2-propoxybenzaldehyde ≥95%
5-Fluoro-2-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD06247718
CAS Number: 19415-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR