4-[2-Bromo-6-fluoro-3-(trifluoromethyl)benzoyl]morpholine ≥97%

Supplier: AOBChem USA

Warning

35463-500MG 35463-1G 35463-5G
77338-326EA 477.92 USD
77338-326 77349-530 77373-762
4-[2-Bromo-6-fluoro-3-(trifluoromethyl)benzoyl]morpholine ≥97%
4-[2-Bromo-6-fluoro-3-(trifluoromethyl)benzoyl]morpholine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrF₄NO₂
MW: 356.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805637
CAS Number: 2271442-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR