1-[2-Bromo-6-fluoro-3-(trifluoromethyl)benzoyl]piperidine ≥97%

Supplier: AOBChem USA

Warning

35462-500MG 35462-1G 35462-5G
77338-836EA 524.27 USD
77338-836 77349-528 77373-760
1-[2-Bromo-6-fluoro-3-(trifluoromethyl)benzoyl]piperidine ≥97%
1-[2-Bromo-6-fluoro-3-(trifluoromethyl)benzoyl]piperidine

Precaución: for Research Use Only

Formula: C₁₃H₁₂BrF₄NO
MW: 354.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805635
CAS Number: 2271442-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR