2-Bromo-6-fluoro-N,N-dimethyl-3-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

35460-500MG 35460-1G 35460-5G
77338-196EA 469.68 USD
77338-196 77349-524 77373-756
2-Bromo-6-fluoro-N,N-dimethyl-3-(trifluoromethyl)benzamide ≥97%
2-Bromo-6-fluoro-N,N-dimethyl-3-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₀H₈BrF₄NO
MW: 314.074 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805638
CAS Number: 2271443-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR