Propan-2-yl 2-Bromo-6-fluoro-3-(trifluoromethyl)benzoate ≥95%

Supplier: AOBChem USA

Warning

35455-500MG 35455-1G 35455-5G
77337-842EA 449.08 USD
77337-842 77349-520 77373-752
Propan-2-yl 2-Bromo-6-fluoro-3-(trifluoromethyl)benzoate ≥95%
Propan-2-yl 2-Bromo-6-fluoro-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₁H₉BrF₄O₂
MW: 329.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805676
CAS Number: 2271443-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR