3-(Benzyloxy)-2,6-dibromopyridine ≥95%

Supplier: AOBChem USA

Warning

35454-1G 35454-5G 35454-25G
77341-084EA 70.04 USD
77341-084 77362-180 77373-750
3-(Benzyloxy)-2,6-dibromopyridine ≥95%
3-(Benzyloxy)-2,6-dibromopyridine

Precaución: for Research Use Only

Formula: C₁₂H₉Br₂NO
MW: 343.014 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564031
CAS Number: 65017-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR