2,6-Dibromo-3-isopropoxypyridine ≥97%

Supplier: AOBChem USA

Warning

35453-1G 35453-5G 35453-25G
77340-642EA 60.77 USD
77340-642 77362-178 77373-748
2,6-Dibromo-3-isopropoxypyridine ≥97%
2,6-Dibromo-3-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₉Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD34564030
CAS Number: 2734774-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR