2,6-Dibromo-3-ethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

35452-1G 35452-5G 35452-25G
77340-640EA 60.77 USD
77340-640 77362-176 77373-746
2,6-Dibromo-3-ethoxypyridine ≥97%
2,6-Dibromo-3-ethoxypyridine

Precaución: for Research Use Only

Formula: C₇H₇Br₂NO
Storage Temperature: Ambient
MDL Number: MFCD00234290
CAS Number: 952569-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR