Ethyl 3-bromo-2-chloro-5-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

35432-500MG 35432-1G 35432-25G 35432-5G
77325-838EA 304.88 USD
77325-838 77349-514 77385-172 77373-744
Ethyl 3-bromo-2-chloro-5-methylbenzoate ≥95%
Ethyl 3-bromo-2-chloro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrClO₂
MW: 277.542 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902590
CAS Number: 2121855-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR