3,5-Dibromo-4-methylphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

35428-500MG 35428-1G 35428-5G
77325-834EA 325.48 USD
77325-834 77349-510 77373-740
3,5-Dibromo-4-methylphenylboronic acid pinacol ester ≥95%
3,5-Dibromo-4-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBr₂O₂
MW: 375.892 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22494363
CAS Number: 1236348-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR