4-Chloro-3-ethoxybenzoic acid ethyl ester ≥97%

Supplier: AOBChem USA

Warning

35415-500MG 35415-1G 35415-5G
77337-552EA 421.27 USD
77337-552 77349-508 77373-738
4-Chloro-3-ethoxybenzoic acid ethyl ester ≥97%
4-Chloro-3-ethoxybenzoic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClO₃
MW: 228.672 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19440068
CAS Number: 909853-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR