4-Bromo-3-chloro-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

35412-500MG 35412-1G 35412-5G 35412-25G
77325-832EA 177.16 USD
77325-832 77346-588 77362-170 77385-170
4-Bromo-3-chloro-N,N-dimethylbenzamide ≥97%
4-Bromo-3-chloro-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrClNO
MW: 262.531 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24020547
CAS Number: 1369900-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR