5-Bromo-2-fluoro-3-methoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

35402-500MG 35402-1G 35402-5G 35402-25G
77325-830EA 196.73 USD
77325-830 77349-506 77362-168 77385-168
5-Bromo-2-fluoro-3-methoxyphenylboronic acid pinacol ester ≥97%
5-Bromo-2-fluoro-3-methoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrFO₃
Storage Temperature: Ambient
MDL Number: MFCD28405317
CAS Number: 1638847-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR