2,3-Dibromo-6-fluorobenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

35399-500MG 35399-1G 35399-5G
77337-054EA 407.88 USD
77337-054 77349-504 77373-736
2,3-Dibromo-6-fluorobenzyl alcohol ≥95%
2,3-Dibromo-6-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅Br₂FO
Storage Temperature: Ambient
MDL Number: MFCD28784711
CAS Number: 1806294-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR