Ethyl 5-bromo-2-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

35392-500MG 35392-1G 35392-5G 35392-25G
77325-828EA 220.42 USD
77325-828 77349-502 77362-164 77385-166
Ethyl 5-bromo-2-fluoro-3-methoxybenzoate ≥97%
Ethyl 5-bromo-2-fluoro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805971
CAS Number: 2112625-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR