Ethyl 3,5-dibromothiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

35391-500MG 35391-1G 35391-5G 35391-25G
77325-826EA 238.96 USD
77325-826 77349-500 77362-162 77385-164
Ethyl 3,5-dibromothiophene-2-carboxylate ≥97%
Ethyl 3,5-dibromothiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆Br₂O₂S
MW: 313.994 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26387207
CAS Number: 62224-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR