1-(4-Iodo-2-pyridinyl)-4-methyl-piperazine ≥97%

Supplier: AOBChem USA

Warning

35372-500MG 35372-1G 35372-5G 35372-25G
77325-822EA 261.62 USD
77325-822 77349-496 77362-158 77385-160
1-(4-Iodo-2-pyridinyl)-4-methyl-piperazine ≥97%
1-(4-Iodo-2-pyridinyl)-4-methyl-piperazine

Precaución: for Research Use Only

Formula: C₁₀H₁₄IN₃
MW: 303.143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34550424
CAS Number: 2072806-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR