4'-Chloro-2,6-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥95%
Supplier: AOBChem USA
|
Warning
|
35350-500MG
35350-1G
35350-5G
35350-25G
77325-820EA
264.71
USD
77325-820
77349-492
77362-156
77385-158
4'-Chloro-2,6-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥95%
4'-Chloro-2,6-dimethyl-[1,1'-biphenyl]-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₅H₁₃ClO Storage Temperature: Ambient |
MDL Number:
MFCD31806024 CAS Number: 1462937-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...