3'-Chloro-2,6-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

35349-500MG 35349-1G 35349-5G 35349-25G
77325-818EA 204.97 USD
77325-818 77349-490 77362-154 77385-156
3'-Chloro-2,6-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3'-Chloro-2,6-dimethyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO
Storage Temperature: Ambient
MDL Number: MFCD31805796
CAS Number: 2271442-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR