6-Methyl-1H-indazole-5-boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
35306-500MG
35306-1G
35306-5G
35306-25G
77325-816EA
90.64
USD
77325-816
77342-428
77362-152
77373-730
6-Methyl-1H-indazole-5-boronic acid ≥97%
6-Methyl-1H-indazole-5-boronic acid
Precaución: for Research Use Only
Formula:
C₈H₉BN₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD12028645 CAS Number: 1310405-35-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...