2-(Benzyloxy)-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

35273-500MG 35273-1G 35273-5G
77340-276EA 873.44 USD
77340-276 77349-482 77373-720
2-(Benzyloxy)-4-methylphenol ≥97%
2-(Benzyloxy)-4-methylphenol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD23140374
CAS Number: 1025708-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR