Ethyl 2-bromo-6-fluoro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

35266-500MG 35266-1G 35266-5G
77337-748EA 440.84 USD
77337-748 77349-478 77373-716
Ethyl 2-bromo-6-fluoro-3-(trifluoromethyl)benzoate ≥97%
Ethyl 2-bromo-6-fluoro-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₇BrF₄O₂
MW: 315.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805677
CAS Number: 2271442-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR