4-(5,5-Dimethyl-1,3,2-Dioxaborinan-2-yl)-3-methyl-benzonitrile ≥97%

Supplier: AOBChem USA

Warning

35264-500MG 35264-1G 35264-5G 35264-25G
77325-810EA 218.36 USD
77325-810 77349-476 77362-148 77385-152
4-(5,5-Dimethyl-1,3,2-Dioxaborinan-2-yl)-3-methyl-benzonitrile ≥97%
4-(5,5-Dimethyl-1,3,2-Dioxaborinan-2-yl)-3-methyl-benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₂
MW: 229.083 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805791
CAS Number: 2231089-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR