(4-Ethoxy-3,5-dimethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

35247-500MG 35247-1G 35247-5G 35247-25G
77325-804EA 108.15 USD
77325-804 77343-162 77362-146 77373-710
(4-Ethoxy-3,5-dimethylphenyl)methanol ≥97%
(4-Ethoxy-3,5-dimethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O₂
MW: 180.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11188447
CAS Number: 1038733-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR