2,3-Difluoro-6-(phenylmethoxy)benzoic acid methyl ester ≥95%

Supplier: AOBChem USA

Warning

35246-500MG 35246-1G 35246-5G 35246-25G
77325-802EA 259.56 USD
77325-802 77349-470 77362-144 77385-148
2,3-Difluoro-6-(phenylmethoxy)benzoic acid methyl ester ≥95%
2,3-Difluoro-6-(phenylmethoxy)benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₅H₁₂F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD31805790
CAS Number: 2271442-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR