2,3-Difluoro-6-(phenylmethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

35245-500MG 35245-1G 35245-5G 35245-25G
77325-800EA 208.06 USD
77325-800 77349-468 77362-142 77385-146
2,3-Difluoro-6-(phenylmethoxy)benzoic acid ≥97%
2,3-Difluoro-6-(phenylmethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD31805789
CAS Number: 2271442-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR