1-Bromo-2-ethoxy-3-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

35235-500MG 35235-1G 35235-5G 35235-25G
77325-796EA 211.15 USD
77325-796 77349-464 77362-138 77385-144
1-Bromo-2-ethoxy-3-methoxybenzene ≥95%
1-Bromo-2-ethoxy-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
MW: 231.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805960
CAS Number: 1269479-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR