2,6-Dichloro-N,N-dimethylaniline ≥95%

Supplier: AOBChem USA

Warning

35222-500MG 35222-1G 35222-5G 35222-25G
77325-794EA 140.08 USD
77325-794 77344-704 77362-136 77385-142
2,6-Dichloro-N,N-dimethylaniline ≥95%
2,6-Dichloro-N,N-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₉Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD29092485
CAS Number: 56961-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR