4-Ethoxy-3,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

35220-500MG 35220-1G 35220-5G 35220-25G
77325-792EA 188.49 USD
77325-792 77349-462 77362-134 77385-140
4-Ethoxy-3,5-dimethylbenzoic acid ≥97%
4-Ethoxy-3,5-dimethylbenzoic acid
Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD11188420
CAS Number: 91970-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR