4-Bromo-2-fluoro-6-methylphenol ≥95%

Supplier: AOBChem USA

Warning

35214-500MG 35214-1G 35214-5G
77325-790EA 352.26 USD
77325-790 77349-460 77373-704
4-Bromo-2-fluoro-6-methylphenol ≥95%
4-Bromo-2-fluoro-6-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
Storage Temperature: Ambient
MDL Number: MFCD24715362
CAS Number: 945557-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR