Ethyl 6-bromo-3-chloro-2-fluorobenzoate ≥95%

Supplier: AOBChem USA

Warning

35211-500MG 35211-1G 35211-5G
77335-670EA 363.59 USD
77335-670 77349-458 77373-702
Ethyl 6-bromo-3-chloro-2-fluorobenzoate ≥95%
Ethyl 6-bromo-3-chloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28142058
CAS Number: 1804897-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR