(3-Chloro-2-fluoro-6-methoxyphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

35173-500MG 35173-1G 35173-5G
77338-956EA 536.63 USD
77338-956 77349-456 77373-700
(3-Chloro-2-fluoro-6-methoxyphenyl)boronic acid ≥97%
(3-Chloro-2-fluoro-6-methoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClFO₃
MW: 204.391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32710934
CAS Number: 2468229-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR