2,3-Difluoro-1,4-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

35162-1G 35162-5G 35162-25G
77340-988EA 66.95 USD
77340-988 77362-132 77373-698
2,3-Difluoro-1,4-dimethoxybenzene ≥97%
2,3-Difluoro-1,4-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
MW: 174.145 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28788791
CAS Number: 845295-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR