Benzyl 2-(benzyloxy)-3-bromobenzoate ≥95%

Supplier: AOBChem USA

Warning

35159-500MG 35159-1G 35159-25G 35159-5G
77325-788EA 284.28 USD
77325-788 77349-454 77385-138 77373-696
Benzyl 2-(benzyloxy)-3-bromobenzoate ≥95%
Benzyl 2-(benzyloxy)-3-bromobenzoate

Precaución: for Research Use Only

Formula: C₂₁H₁₇BrO₃
MW: 397.262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707005
CAS Number: 2385018-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR