3-Bromo-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

35153-500MG 35153-1G 35153-5G 35153-25G
77325-784EA 128.75 USD
77325-784 77344-300 77362-128 77373-694
3-Bromo-2-isopropoxybenzoic acid ≥97%
3-Bromo-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₃
MW: 259.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14656685
CAS Number: 1247577-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR