3-Bromo-2-propoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

35149-500MG 35149-1G 35149-5G
77339-348EA 581.95 USD
77339-348 77349-448 77373-690
3-Bromo-2-propoxybenzaldehyde ≥95%
3-Bromo-2-propoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14656659
CAS Number: 1247906-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR