Ethyl 3,6-dibromo-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

35147-500MG 35147-1G 35147-5G 35147-25G
77325-782EA 233.81 USD
77325-782 77349-446 77362-126 77385-134
Ethyl 3,6-dibromo-2-fluorobenzoate ≥97%
Ethyl 3,6-dibromo-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇Br₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD14698370
CAS Number: 1214347-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR