Ethyl-2-chloro-4-iodonicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-chloro-4-iodopyridine-3-carboxylate

35146-500MG 35146-1G 35146-5G 35146-25G
77325-780EA 106.09 USD
77325-780 77343-114 77362-124 77373-688
Ethyl-2-chloro-4-iodonicotinate ≥97%
Ethyl-2-chloro-4-iodonicotinate

Precaución: for Research Use Only

Formula: C₈H₇ClINO₂
MW: 311.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781133
CAS Number: 219727-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR