Methyl 3,6-dibromo-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

35142-500MG 35142-1G 35142-5G
77325-778EA 311.06 USD
77325-778 77349-444 77373-686
Methyl 3,6-dibromo-2-fluorobenzoate ≥97%
Methyl 3,6-dibromo-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅Br₂FO₂
MW: 311.931 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698153
CAS Number: 1214329-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR