3-Bromo-2-ethoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

35113-500MG 35113-1G 35113-5G 35113-25G
77325-774EA 201.88 USD
77325-774 77349-440 77362-122 77385-130
3-Bromo-2-ethoxyphenylboronic acid ≥95%
3-Bromo-2-ethoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BBrO₃
MW: 244.878 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18968534
CAS Number: 352525-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR