(4-Bromo-3-fluoro-2-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

35107-500MG 35107-1G 35107-5G
77338-552EA 498.52 USD
77338-552 77349-438 77373-682
(4-Bromo-3-fluoro-2-methylphenyl)methanol ≥97%
(4-Bromo-3-fluoro-2-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805992
CAS Number: 2012740-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR